Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001242854.1 | NP_001229783.1 | 5 | 722 | |
NM_001242855.1 | NP_001229784.1 | 4 | 482 | ||
NM_001242856.1 | NP_001229785.1 | 3 | 363 | ||
NM_012402.3 | NP_036534.1 | 5 | 583 | ||
XM_005252840.4 | XP_005252897.1 | 5 | 487 | ||
GenBank mRNA | AK026390.1 | - | 5 | 415 | |
AK300855.1 | - | 5 | 583 | ||
AK301856.1 | - | 4 | 437 | ||
AK301972.1 | - | 5 | 524 | ||
AK304512.1 | - | 3 | 230 | ||
AK311004.1 | - | 1 | 192 | ||
AK311086.1 | - | 4 | 1065 | ||
BC000392.2 | - | 5 | 405 | 2821666 | |
BM928413.1 | - | 550 | |||
U52522.1 | - | 5 | 399 | ||
X97567.1 | - | 3 | 218 |