Gene Transcripts

Gene Symbol
ABT1
Entrez Gene ID
29777
Gene Name
activator of basal transcription 1
Gene Aliases
hABT1
Chromosome Location
Chr. 6: 26596943 - 26600050 on Build GRCh38
UniGene ID
Hs.254406
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013375.3 NP_037507.1 3 974
GenBank mRNA AB027258.1 - 3 954
BC048812.1 - 3 972 5574755
BC066313.1 - 3 964 5272771

View Full Product Details