Gene Transcripts

Gene Symbol
VPREB3
Entrez Gene ID
29802
Gene Name
pre-B lymphocyte 3
Gene Aliases
8HS20, N27C7-2
Chromosome Location
Chr. 22: 23752743 - 23754443 on Build GRCh38
UniGene ID
Hs.136713
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013378.2 NP_037510.1 2 418
GenBank mRNA AF163825.1 - 2 380
AK312098.1 - 2 373
AY359000.1 - 2 418
BC020666.1 - 2 443 4772032
BQ899182.1 - 2 351
CR456610.1 - 2 367

View Full Product Details