Gene Transcripts

Gene Symbol
NRBP1
Entrez Gene ID
29959
Gene Name
nuclear receptor binding protein 1
Gene Aliases
BCON3, MADM, MUDPNP, NRBP
Chromosome Location
Chr. 2: 27427790 - 27442259 on Build GRCh38
UniGene ID
Hs.515876
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321357.1 NP_001308286.1 17 2270
NM_001321358.1 NP_001308287.1 17 1667
NM_001321359.1 NP_001308288.1 17 1662
NM_001321361.1 NP_001308290.1 17 1664
NM_001321362.1 NP_001308291.1 16 1640
NM_001321363.1 NP_001308292.1 16 1547
NM_013392.3 NP_037524.1 16 1638
GenBank mRNA AF113249.1 - 16 1550
AK001946.1 - 16 1528
AK027538.1 - 16 1546
AK092860.1 - 9 1948
AK122664.1 - 17 2270
AK130619.1 - 9 682
AK223136.1 - 16 1534
AK301345.1 - 17 1481
AL136682.1 - 16 1525
BC001221.1 - 16 1552 3534543
BQ225378.1 - 2 87

View Full Product Details