Gene Transcripts

Gene Symbol
MKRN2
Entrez Gene ID
23609
Gene Name
makorin ring finger protein 2
Gene Aliases
HSPC070, RNF62
Chromosome Location
Chr. 3: 12557014 - 12583713 on Build GRCh38
UniGene ID
Hs.591666
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271707.1 NP_001258636.1 7 2187
NM_014160.4 NP_054879.3 8 2316
GenBank mRNA AF302084.1 - 8 2230
AK091318.1 - 8 2261
AL834512.1 - 5 1838
BC001799.2 - 6 2025
BC015715.1 - 8 2258 3916199

View Full Product Details