Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014321.3 | NP_055136.1 | 7 | 716 | |
NR_037620.1 | 7 | 775 | |||
GenBank mRNA | AF139658.1 | - | 7 | 704 | |
AK021946.1 | - | 7 | 677 | ||
AK024019.1 | - | 7 | 710 | ||
BC039032.1 | - | 7 | 696 | 5577396 | |
BC063565.1 | - | 7 | 685 | 3460593 |