Gene Transcripts
- Gene Symbol
- GHITM
- Entrez Gene ID
- 27069
- Gene Name
- growth hormone inducible transmembrane protein
- Gene Aliases
- DERP2, HSPC282, MICS1, My021, PTD010, TMBIM5
- Chromosome Location
- Chr. 10: 84139429 - 84153555 on Build GRCh38
- UniGene ID
- Hs.352656
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_014394.2 |
NP_055209.2
|
2, 3 |
333 |
|
GenBank mRNA |
AB009685.1 |
- |
1, 2 |
129 |
|
AF060923.1 |
- |
2, 3 |
207 |
|
AF078863.1 |
- |
2, 3 |
258 |
|
AF131820.1 |
- |
2, 3 |
260 |
|
AF161400.1 |
- |
2, 3 |
242 |
|
AK098770.1 |
- |
2, 3 |
206 |
|
AK130094.1 |
- |
2, 3 |
236 |
|
AK130097.1 |
- |
2, 3 |
201 |
|
AK225318.1 |
- |
2, 3 |
210 |
|
AK292864.1 |
- |
2, 3 |
201 |
|
AK297960.1 |
- |
2, 3 |
200 |
|
AK299937.1 |
- |
|
239 |
|
AL136713.1 |
- |
2, 3 |
243 |
|
AY358824.1 |
- |
2, 3 |
208 |
|
BC010354.1 |
- |
2, 3 |
204 |
4093037 |
BC022872.1 |
- |
2, 3 |
215 |
|
CR533519.1 |
- |
1, 2 |
129 |
|
View Full Product Details