Gene Transcripts

Gene Symbol
TOR1B
Entrez Gene ID
27348
Gene Name
torsin family 1 member B
Gene Aliases
DQ1
Chromosome Location
Chr. 9: 129803143 - 129811424 on Build GRCh38
UniGene ID
Hs.252682
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317893.1 NP_001304822.1 3 651
NM_014506.2 NP_055321.1 3 651
NR_133948.1 3 792
NR_134113.1 3 792
XM_005251927.3 XP_005251984.1 3 646
GenBank mRNA AF007872.1 - 2 372
AF317129.1 - 3 641
AI568476.1 - 2 348
BC015578.2 - 3 596 4589479
CN262366.1 - 2 441

View Full Product Details