Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003568.2 | NP_003559.2 | 13 | 1416 | |
GenBank mRNA | AF230929.1 | - | 13 | 1172 | |
AJ009985.1 | - | 13 | 1361 | ||
AK314926.1 | - | 12 | 1269 | ||
BC005830.2 | - | 12 | 1072 | 2959488 | |
CR536481.1 | - | 11 | 925 |