Gene Transcripts

Gene Symbol
TIPARP
Entrez Gene ID
25976
Gene Name
TCDD inducible poly(ADP-ribose) polymerase
Gene Aliases
ARTD14, PARP7, pART14
Chromosome Location
Chr. 3: 156674416 - 156706770 on Build GRCh38
UniGene ID
Hs.744050
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001184717.1 NP_001171646.1 2 766
NM_001184718.1 NP_001171647.1 2 331
NM_015508.4 NP_056323.2 2 681
GenBank mRNA AK097785.1 - 2 510
AK291261.1 - 2 516
BC034397.1 - 2 331
BC050350.1 - 2 488 5264807
BX537965.1 - 2 508
CR749647.1 - 2 767

View Full Product Details