Gene Transcripts
- Gene Symbol
- CFAP61
- Entrez Gene ID
- 26074
- Gene Name
- cilia and flagella associated protein 61
- Gene Aliases
- C20orf26, CaM-IP3, dJ1002M8.3, dJ1178H5.4
- Chromosome Location
- Chr. 20: 20052514 - 20360714 on Build GRCh38
- UniGene ID
- Hs.729168
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001167816.1 |
NP_001161288.1
|
4, 5 |
450 |
|
NM_015585.3 |
NP_056400.3
|
5, 6 |
556 |
|
XM_005260688.3 |
XP_005260745.1
|
5, 6 |
519 |
|
XM_005260689.2 |
XP_005260746.1
|
5, 6 |
519 |
|
XM_005260690.3 |
XP_005260747.1
|
5, 6 |
519 |
|
XM_005260691.3 |
XP_005260748.1
|
5, 6 |
519 |
|
XM_005260692.3 |
XP_005260749.1
|
5, 6 |
519 |
|
XM_005260693.3 |
XP_005260750.1
|
5, 6 |
519 |
|
XM_005260697.1 |
XP_005260754.1
|
5, 6 |
519 |
|
XM_011529211.1 |
XP_011527513.1
|
4, 5 |
377 |
|
XM_011529212.2 |
XP_011527514.1
|
5, 6 |
519 |
|
XM_011529213.1 |
XP_011527515.1
|
5, 6 |
519 |
|
XM_017027786.1 |
XP_016883275.1
|
5, 6 |
519 |
|
XM_017027787.1 |
XP_016883276.1
|
5, 6 |
519 |
|
XM_017027788.1 |
XP_016883277.1
|
5, 6 |
519 |
|
XM_017027789.1 |
XP_016883278.1
|
5, 6 |
519 |
|
XM_017027790.1 |
XP_016883279.1
|
5, 6 |
519 |
|
XM_017027795.1 |
XP_016883284.1
|
5, 6 |
519 |
|
GenBank mRNA |
AK304174.1 |
- |
5, 6 |
519 |
|
AK307817.1 |
- |
4, 5 |
377 |
|
AL117439.1 |
- |
4, 5 |
388 |
|
BC028708.2 |
- |
5, 6 |
518 |
4825941 |
View Full Product Details