Gene Transcripts

Gene Symbol
AIG1
Entrez Gene ID
51390
Gene Name
androgen induced 1
Gene Aliases
AIG-1, dJ95L4.1
Chromosome Location
Chr. 6: 143059363 - 143343594 on Build GRCh38
UniGene ID
Hs.567501
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286587.1 NP_001273516.1 5 442
NM_016108.3 NP_057192.2 4 499
XM_011535883.2 XP_011534185.1 4 480
XM_011535884.2 XP_011534186.1 4 482
XM_017010927.1 XP_016866416.1 4 498
GenBank mRNA AF151861.1 - 5 445
AF153605.1 - 4 442
AJ420480.1 - 2 115
AK001347.1 - 4 440
AK222475.1 - 4 461
BC025278.1 - 4 430 4859216
BF725860.1 - 3 332
BX538067.1 - 3 202

View Full Product Details