Gene Transcripts

Gene Symbol
SUCO
Entrez Gene ID
51430
Gene Name
SUN domain containing ossification factor
Gene Aliases
C1orf9, CH1, OPT, SLP1
Chromosome Location
Chr. 1: 172532349 - 172611833 on Build GRCh38
UniGene ID
Hs.204559
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282751.1 NP_001269680.1 16 2811
NM_014283.4 NP_055098.1 18 2971
NM_016227.3 NP_057311.3 17 3230
XM_006711374.2 XP_006711437.1 17 3031
XM_006711375.2 XP_006711438.1 17 2941
XM_006711376.1 XP_006711439.1 16 2920
GenBank mRNA AF097535.1 - 17 3230
AJ250075.1 - 18 2969
AK302949.1 - 18 2971
AL035291.1 - 18 2872
BC137125.1 - 18 2757 9021122
BC144487.1 - 16 2597

View Full Product Details