Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018094.4 | NP_060564.2 | 1 | 551 | |
GenBank mRNA | AJ251548.1 | - | 1 | 309 | |
AK001303.1 | - | 1 | 489 | ||
AK023155.1 | - | 1 | 495 | ||
AK225671.1 | - | 1 | 516 | ||
AK298868.1 | - | 2 | 245 | ||
BC036077.1 | - | 1 | 505 | 5313878 | |
DB173006.1 | - | 1 | 552 |