Gene Transcripts

Gene Symbol
DEF6
Entrez Gene ID
50619
Gene Name
DEF6, guanine nucleotide exchange factor
Gene Aliases
IBP, SLAT, SWAP70L
Chromosome Location
Chr. 6: 35297818 - 35321771 on Build GRCh38
UniGene ID
Hs.15476
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022047.3 NP_071330.3 11 1932
GenBank mRNA AK093613.1 - 10 1644
AL110182.1 - 1 134
AY241694.1 - 11 1932
BC017504.1 - 11 1898

View Full Product Details