Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005843.5 | NP_005834.4 | 6 | 856 | |
GenBank mRNA | AF042273.1 | - | 6 | 615 | |
AF042274.1 | - | 6 | 615 | ||
AK292265.1 | - | 6 | 551 | ||
AK292847.1 | - | 6 | 593 | ||
AK310863.1 | - | 6 | 593 | ||
AL133600.1 | - | 6 | 618 | ||
BC028740.2 | - | 6 | 856 | 5260998 |