Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001130690.2 | NP_001124162.1 | 4 | 364 | |
NM_006661.3 | NP_006652.1 | 5 | 492 | ||
XM_006715321.3 | XP_006715384.1 | 4 | 1689 | ||
XM_011535387.2 | XP_011533689.2 | 6 | 2484 | ||
XM_011535388.2 | XP_011533690.1 | 5 | 1817 | ||
XM_011535393.2 | XP_011533695.1 | 3 | 137 | ||
XM_017010194.1 | XP_016865683.1 | 6 | 2484 | ||
XM_017010195.1 | XP_016865684.1 | 4 | 237 | ||
XM_017010196.1 | XP_016865685.1 | 6 | 1935 | ||
XM_017010197.1 | XP_016865686.1 | 6 | 2484 | ||
GenBank mRNA | AB020593.1 | - | 5 | 497 | |
AB026816.1 | - | 4 | 297 | ||
AF127479.1 | - | 4 | 313 | ||
AF127480.1 | - | 4 | 333 | ||
BC104858.1 | - | 4 | 284 | 8143861 | |
BC104860.1 | - | 4 | 284 | 8143863 | |
BC143297.1 | - | 5 | 403 | ||
BC143301.1 | - | 5 | 403 | ||
CR536567.1 | - | 4 | 247 | ||
DR000181.1 | - | 4 | 575 | ||
KU232567.1 | - | 4 | 735 |