Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018425.3 | NP_060895.1 | 7 | 1262 | |
GenBank mRNA | AJ303098.1 | - | 7 | 1262 | |
AL353952.1 | - | 5 | 543 | ||
BC003167.1 | - | 7 | 1234 | 2905670 | |
BT007330.1 | - | 7 | 1205 |