Gene Transcripts

Gene Symbol
HCP5
Entrez Gene ID
10866
Gene Name
HLA complex P5 (non-protein coding)
Gene Aliases
6S2650E, D6S2650E, P5-1
Chromosome Location
Chr. 6: 31463180 - 31465809 on Build GRCh38
UniGene ID
Hs.691948
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_040662.1 2 534
GenBank mRNA AK093953.1 - 3 448
AK290875.1 - 2 529
BC106759.2 - 2 480 40024782
BC113853.1 - 1 571
BC114462.1 - 2 480
BX640979.1 - 2 532
DA501560.1 - 2 535
L06175.1 - 2 530

View Full Product Details