Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001261824.1 | NP_001248753.1 | 3 | 491 | |
NM_001320795.1 | NP_001307724.1 | 3 | 679 | ||
NM_003720.3 | NP_003711.1 | 3 | 491 | ||
NM_203433.2 | NP_982257.1 | 3 | 491 | ||
NR_049728.1 | 2 | 384 | |||
XM_017028488.1 | XP_016883977.1 | 3 | 405 | ||
GenBank mRNA | AB451468.1 | - | 3 | 362 | |
AF417108.1 | - | 2 | 255 | ||
AJ006291.1 | - | 3 | 425 | ||
AK001455.1 | - | 3 | 453 | ||
AK308614.1 | - | 2 | 362 | ||
AK315406.1 | - | 3 | 393 | ||
AY463963.1 | - | 3 | 362 | ||
BC003619.2 | - | 3 | 408 | 2961575 | |
BC010424.1 | - | 3 | 400 | 3343732 | |
BC011755.2 | - | 3 | 408 | 3531528 | |
BC012809.2 | - | 3 | 408 | 2961575 | |
BM460414.1 | - | 3 | 384 | ||
BM462074.1 | - | 3 | 423 | ||
BU543452.1 | - | 3 | 442 | ||
CB160221.1 | - | 3 | 353 | ||
CR541821.1 | - | 3 | 362 | ||
CR541852.1 | - | 3 | 362 |