Gene Transcripts

Gene Symbol
RPP14
Entrez Gene ID
11102
Gene Name
ribonuclease P/MRP subunit p14
Gene Aliases
HsHTD2, P14
Chromosome Location
Chr. 3: 58306245 - 58322642 on Build GRCh38
UniGene ID
Hs.446320
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098783.2 NP_001092253.1 6 1258
NM_007042.4 NP_008973.1 6 1024
NR_049755.1 5 1160
NR_049756.1 6 1024
NR_049757.1 5 926
NR_049758.1 4 841
XM_011533313.2 XP_011531615.1 6 1058
XM_017005641.1 XP_016861130.1 6 1021
GenBank mRNA AF001175.1 - 6 1000
AK311396.1 - 5 885
BC002441.1 - 6 972 3347003
BC007342.1 - 6 972 3616325
BC012017.1 - 6 985 4708724

View Full Product Details