Gene Transcripts

Gene Symbol
PXMP4
Entrez Gene ID
11264
Gene Name
peroxisomal membrane protein 4
Gene Aliases
PMP24
Chromosome Location
Chr. 20: 33702744 - 33720330 on Build GRCh38
UniGene ID
Hs.654857
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007238.4 NP_009169.3 4 970
NM_183397.2 NP_899634.1 3 771
GenBank mRNA AF072864.1 - 4 892
AK023879.1 - 4 888
AK297018.1 - 3 760
AL136648.1 - 4 941
BC001147.1 - 4 871 3533572
BQ278083.1 - 3 739

View Full Product Details