Gene Transcripts

Gene Symbol
TMC6
Entrez Gene ID
11322
Gene Name
transmembrane channel like 6
Gene Aliases
EV1, EVER1, EVIN1, LAK-4P
Chromosome Location
Chr. 17: 78108775 - 78132427 on Build GRCh38
UniGene ID
Hs.632227
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127198.2 NP_001120670.1 14 2050
NM_001321185.1 NP_001308114.1 14 2003
NM_007267.7 NP_009198.4 14 1954
XM_005256995.1 XP_005257052.1 14 2079
XM_011524255.1 XP_011522557.1 13 1981
XM_011524256.1 XP_011522558.1 13 1855
XM_011524257.2 XP_011522559.1 8 1099
XM_017024107.1 XP_016879596.1 13 1848
XM_017024108.1 XP_016879597.1 13 1801
XM_017024109.1 XP_016879598.1 12 1770
GenBank mRNA AB002405.2 - 6 762
AK074065.1 - 14 1953
AK310734.1 - 13 2374
AY057379.1 - 13 1793
AY099356.1 - 15 1952
AY236496.1 - 13 1919
BC018346.1 - 7 918
BC023597.2 - 14 1934 4566166
BC035648.1 - 7 974 5575949
DQ104440.1 - 4 530

View Full Product Details