Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001012478.1 | NP_001012496.1 | 2, 3 | 1241 | |
NM_007279.2 | NP_009210.1 | 2, 3 | 1241 | ||
XM_006722994.2 | XP_006723057.1 | 3, 4 | 1279 | ||
XM_011526410.1 | XP_011524712.1 | 3, 4 | 1279 | ||
GenBank mRNA | AB451261.1 | - | 2, 3 | 186 | |
AB451387.1 | - | 2, 3 | 186 | ||
BC008740.2 | - | 2, 3 | 245 | 3346087 | |
BC030574.2 | - | 2, 3 | 1145 | 3687471 | |
X64044.1 | - | 2, 3 | 226 |