Gene Transcripts
- Gene Symbol
- ETHE1
- Entrez Gene ID
- 23474
- Gene Name
- ETHE1, persulfide dioxygenase
- Gene Aliases
- HSCO, YF13H12
- Chromosome Location
- Chr. 19: 43506719 - 43527256 on Build GRCh38
- UniGene ID
- Hs.7486
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001320867.1 |
NP_001307796.1
|
5, 6 |
639 |
|
NM_001320868.1 |
NP_001307797.1
|
4, 5 |
523 |
|
NM_001320869.1 |
NP_001307798.1
|
3, 4 |
378 |
|
NM_014297.4 |
NP_055112.2
|
5, 6 |
672 |
|
XM_005258687.3 |
XP_005258744.1
|
5, 6 |
769 |
|
GenBank mRNA |
AK309880.1 |
- |
5, 6 |
617 |
|
AK315346.1 |
- |
5, 6 |
635 |
|
BC008250.1 |
- |
5, 6 |
622 |
3872059 |
BG706505.1 |
- |
4, 5 |
472 |
|
D83198.2 |
- |
5, 6 |
661 |
|
KU178565.1 |
- |
5, 6 |
593 |
|
KU178566.1 |
- |
5, 6 |
560 |
|
KU178567.1 |
- |
3, 4 |
299 |
|
View Full Product Details