Gene Transcripts

Gene Symbol
NLGN4Y
Entrez Gene ID
22829
Gene Name
neuroligin 4, Y-linked
Gene Aliases
Chromosome Location
Chr. Y: 14522578 - 14844945 on Build GRCh38
UniGene ID
Hs.439199
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001206850.1 NP_001193779.1 6 2899
NM_014893.4 NP_055708.3 6 3229
NR_028319.1 6 3451
NR_046355.1 7 3216
XM_006724874.2 XP_006724937.1 7 3461
XM_011531424.2 XP_011529726.1 7 3779
XM_011531425.2 XP_011529727.1 7 3551
XM_011531426.2 XP_011529728.1 7 3512
XM_011531427.1 XP_011529729.1 7 3266
XM_011531428.1 XP_011529730.1 8 3447
XM_011531429.2 XP_011529731.1 7 4062
XM_011531430.2 XP_011529732.1 7 3400
XM_017030034.1 XP_016885523.1 6 3491
XM_017030035.1 XP_016885524.1 6 3412
XM_017030036.1 XP_016885525.1 6 3206
XM_017030037.1 XP_016885526.1 6 3204
XM_017030038.1 XP_016885527.1 6 4002
XM_017030039.1 XP_016885528.1 7 2737
XM_017030040.1 XP_016885529.1 6 2586
GenBank mRNA AB023168.1 - 6 2755
AF376804.1 - 6 3229
BX537428.1 - 6 3452

View Full Product Details