Gene Transcripts

Gene Symbol
PMPCA
Entrez Gene ID
23203
Gene Name
peptidase, mitochondrial processing alpha subunit
Gene Aliases
Alpha-MPP, INPP5E, P-55, SCAR2
Chromosome Location
Chr. 9: 136410573 - 136423761 on Build GRCh38
UniGene ID
Hs.495471
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282944.1 NP_001269873.1 5, 6 643
NM_001282946.1 NP_001269875.1 6, 7 736
NM_015160.2 NP_055975.1 6, 7 738
XM_005266059.4 XP_005266116.1 6, 7 653
XM_011518417.2 XP_011516719.1 2, 3 192
XM_017014543.1 XP_016870032.1 5, 6 599
GenBank mRNA AK296617.1 - 5, 6 555
AK308104.1 - 6, 7 651
BC022949.1 - 6, 7 630
BC033103.1 - 6, 7 647
BC111399.1 - 6, 7 653 4609491
BC132724.1 - 6, 7 677 40146746
BC136599.1 - 6, 7 677 9020589
D21064.1 - 6, 7 651
D50913.2 - 6, 7 651

View Full Product Details