Gene Transcripts

Gene Symbol
TCAP
Entrez Gene ID
8557
Gene Name
titin-cap
Gene Aliases
CMD1N, CMH25, LGMD2G, T-cap, TELE, telethonin
Chromosome Location
Chr. 17: 39665346 - 39666554 on Build GRCh38
UniGene ID
Hs.77628
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003673.3 NP_003664.1 2 770
GenBank mRNA AJ000491.1 - 2 766
AK096328.1 - 2 1929
BC012628.1 - 2 771 4251080
BC013330.1 - 2 771 4273711

View Full Product Details