Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001271734.1 | NP_001258663.1 | 8 | 1326 | |
NM_001271735.1 | NP_001258664.1 | 8 | 1344 | ||
NM_001271736.1 | NP_001258665.1 | 9 | 1479 | ||
NM_016057.2 | NP_057141.1 | 9 | 1395 | ||
NR_073424.1 | 8 | 1303 | |||
GenBank mRNA | AF086911.1 | - | 9 | 1342 | |
AF151878.1 | - | 9 | 1358 | ||
AK025956.1 | - | 9 | 1354 | ||
AK026190.1 | - | 9 | 1354 | ||
BC002849.2 | - | 9 | 1337 | 3637869 |