Gene Transcripts

Gene Symbol
NOL4
Entrez Gene ID
8715
Gene Name
nucleolar protein 4
Gene Aliases
CT125, HRIHFB2255, NOLP
Chromosome Location
Chr. 18: 33851100 - 34224834 on Build GRCh38
UniGene ID
Hs.514795
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001198546.1 NP_001185475.1 3 754
NM_001198547.1 NP_001185476.1 3 643
NM_001198548.1 NP_001185477.1 3 754
NM_001282527.1 NP_001269456.1 4 684
NM_003787.4 NP_003778.2 3 754
XM_006722563.2 XP_006722626.1 4 1511
XM_006722564.2 XP_006722627.1 3 496
XM_006722565.1 XP_006722628.1 4 530
XM_011526237.1 XP_011524539.1 4 1508
XM_011526238.1 XP_011524540.1 3 1120
XM_011526239.2 XP_011524541.1 2 512
XM_017026052.1 XP_016881541.1 2 342
XM_017026053.1 XP_016881542.1 3 494
XM_017026054.1 XP_016881543.1 4 622
XM_017026055.1 XP_016881544.1 4 1161
XM_017026056.1 XP_016881545.1 4 415
XM_017026057.1 XP_016881546.1 3 874
XM_017026058.1 XP_016881547.1 3 233
XM_017026059.1 XP_016881548.1 3 2036
GenBank mRNA AB017800.1 - 3 764
AK091463.1 - 4 533
AK297185.1 - 3 408
AK299850.1 - 3 265
AK312562.1 - 4 281
BC000313.2 - 3 440 2821116
BT006763.1 - 2 114
CR456730.1 - 2 114

View Full Product Details