Gene Transcripts

Gene Symbol
SH2D2A
Entrez Gene ID
9047
Gene Name
SH2 domain containing 2A
Gene Aliases
F2771, SCAP, TSAD, VRAP
Chromosome Location
Chr. 1: 156806238 - 156816853 on Build GRCh38
UniGene ID
Hs.103527
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161441.1 NP_001154913.1 6 828
NM_001161442.1 NP_001154914.1 6 767
NM_001161443.1 NP_001154915.1 5 677
NM_001161444.1 NP_001154916.1 6 798
NM_003975.3 NP_003966.2 6 798
XM_006711615.2 XP_006711678.1 6 816
XM_017002762.1 XP_016858251.1 6 845
XM_017002763.1 XP_016858252.1 6 836
XM_017002764.1 XP_016858253.1 6 783
XM_017002765.1 XP_016858254.1 6 796
XM_017002766.1 XP_016858255.1 6 786
XM_017002767.1 XP_016858256.1 6 851
GenBank mRNA AF051325.1 - 6 720
AF097744.1 - 6 697
AJ000553.1 - 6 744
AK222737.1 - 6 721
AK313706.1 - 6 792
AY763098.1 - 5 677
AY763099.1 - 6 774
AY763100.1 - 6 704
BC012107.1 - 6 760 4636736

View Full Product Details