Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001321135.1 | NP_001308064.1 | 6 | 887 | |
NM_001321136.1 | NP_001308065.1 | 7 | 1106 | ||
NM_001321137.1 | NP_001308066.1 | 7 | 703 | ||
NM_003978.4 | NP_003969.2 | 6 | 887 | ||
NR_135552.1 | 6 | 887 | |||
XM_006720737.2 | XP_006720800.1 | 6 | 619 | ||
XM_011522163.1 | XP_011520465.1 | 7 | 905 | ||
XM_011522164.1 | XP_011520466.1 | 7 | 573 | ||
XM_011522165.2 | XP_011520467.1 | 6 | 428 | ||
XM_011522166.1 | XP_011520468.1 | 7 | 905 | ||
XM_011522167.1 | XP_011520469.1 | 7 | 905 | ||
XM_011522168.2 | XP_011520470.1 | 7 | 905 | ||
XM_011522169.1 | XP_011520471.1 | 7 | 905 | ||
GenBank mRNA | AB451310.1 | - | 6 | 398 | |
AB451440.1 | - | 6 | 398 | ||
AF038602.1 | - | 6 | 398 | ||
AF038603.1 | - | 6 | 398 | ||
AK298733.1 | - | 6 | 640 | ||
AK304052.1 | - | 6 | 676 | ||
AK308462.1 | - | 7 | 703 | ||
AK309856.1 | - | 7 | 1069 | ||
AK310072.1 | - | 7 | 820 | ||
AK316220.1 | - | 6 | 650 | ||
BC008602.1 | - | 6 | 759 | 4180398 | |
KU178458.1 | - | 6 | 398 | ||
KU178459.1 | - | 6 | 398 | ||
U94778.1 | - | 6 | 614 |