Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004760.2 | NP_004751.2 | 7 | 1597 | |
XM_017012792.1 | XP_016868281.1 | 7 | 1409 | ||
GenBank mRNA | AB011420.1 | - | 7 | 1563 | |
AK097170.1 | - | 2 | 2125 | ||
BC023508.2 | - | 7 | 1553 | 3638004 | |
BC047696.1 | - | 7 | 1566 | 5761182 |