Gene Transcripts

Gene Symbol
SNAP29
Entrez Gene ID
9342
Gene Name
synaptosome associated protein 29
Gene Aliases
CEDNIK, SNAP-29
Chromosome Location
Chr. 22: 20859004 - 20891214 on Build GRCh38
UniGene ID
Hs.108002
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004782.3 NP_004773.1 4 704
GenBank mRNA AF115436.1 - 4 671
AK024704.1 - 1 91
BC009715.1 - 4 668 3900895
BT007357.1 - 4 597
CR456582.1 - 4 630

View Full Product Details