Gene Transcripts

Gene Symbol
RGS11
Entrez Gene ID
8786
Gene Name
regulator of G-protein signaling 11
Gene Aliases
RS11
Chromosome Location
Chr. 16: 268301 - 275943 on Build GRCh38
UniGene ID
Hs.65756
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286485.1 NP_001273414.1 12, 13 976
NM_001286486.1 NP_001273415.1 11, 12 844
NM_003834.2 NP_003825.1 13, 14 1020
NM_183337.2 NP_899180.1 13, 14 1009
XM_011522719.2 XP_011521021.1 14, 15 1103
XM_011522720.2 XP_011521022.1 14, 15 1043
GenBank mRNA AB016929.1 - 12, 13 944
AB209582.1 - 5, 6 2917
AF035153.1 - 13, 14 980
AF035154.1 - 13, 14 991
BC141803.1 - 13, 14 984 40147243

View Full Product Details