Gene Transcripts

Gene Symbol
CHAF1A
Entrez Gene ID
10036
Gene Name
chromatin assembly factor 1 subunit A
Gene Aliases
CAF-1, CAF1, CAF1B, CAF1P150, P150
Chromosome Location
Chr. 19: 4402596 - 4448322 on Build GRCh38
UniGene ID
Hs.79018
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005483.2 NP_005474.2 15 3040
XM_011527607.2 XP_011525909.1 14 3050
GenBank mRNA BC067093.1 - 15 3041
BE796132.1 - 3 391
U20979.1 - 15 2926

View Full Product Details