Gene Transcripts

Gene Symbol
ARL4C
Entrez Gene ID
10123
Gene Name
ADP ribosylation factor like GTPase 4C
Gene Aliases
ARL7, LAK
Chromosome Location
Chr. 2: 234493041 - 234497053 on Build GRCh38
UniGene ID
Hs.111554
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005737.3 NP_005728.2 1 1166
GenBank mRNA AB016811.1 - 1 674
AJ579851.1 - 1 717
BC089043.1 - 1 1170 6578402

View Full Product Details