Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005804.3 | NP_005795.2 | 7, 8 | 990 | |
XM_006722606.3 | XP_006722669.1 | 8, 9 | 1115 | ||
XM_011527620.1 | XP_011525922.1 | 7, 8 | 1009 | ||
XM_011527621.2 | XP_011525923.1 | 7, 8 | 934 | ||
GenBank mRNA | AK301847.1 | - | 8, 9 | 1073 | |
AL512698.1 | - | 1, 2 | 1196 | ||
BC001009.2 | - | 7, 8 | 927 | 2820926 | |
BM792110.1 | - | 1, 2 | 123 | ||
U90426.1 | - | 7, 8 | 967 |