Gene Transcripts

Gene Symbol
LAPTM4A
Entrez Gene ID
9741
Gene Name
lysosomal protein transmembrane 4 alpha
Gene Aliases
HUMORF13, LAPTM4, MBNT, Mtrp
Chromosome Location
Chr. 2: 20032650 - 20052028 on Build GRCh38
UniGene ID
Hs.467807
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014713.4 NP_055528.1 6 1092
GenBank mRNA AK129875.1 - 6 742
AY359028.1 - 6 708
BC000421.2 - 6 669 2964450
BC003158.1 - 6 732 2958870
BC093075.1 - 6 692 30402963
CR456758.1 - 6 584
D14696.1 - 6 730

View Full Product Details