Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003958.3 | NP_003949.1 | 2 | 328 | |
NM_183078.2 | NP_898901.1 | 2 | 328 | ||
NR_046399.1 | 2 | 328 | |||
XM_006715241.2 | XP_006715304.1 | 2 | 328 | ||
XM_006715242.2 | XP_006715305.1 | 2 | 328 | ||
XM_017011462.1 | XP_016866951.1 | 2 | 179 | ||
XM_017011463.1 | XP_016866952.1 | 2 | 184 | ||
XM_017011464.1 | XP_016866953.1 | 2 | 227 | ||
GenBank mRNA | AB014546.1 | - | 2 | 247 | |
AF334675.1 | - | 2 | 135 | ||
AK022075.1 | - | 2 | 229 | ||
AK222765.1 | - | 2 | 246 | ||
AK298319.1 | - | 2 | 246 | ||
AK308840.1 | - | 2 | 235 | ||
AL711279.1 | - | 2 | 286 | ||
BC007517.2 | - | 2 | 257 | 3028746 | |
BT007446.1 | - | 2 | 135 | ||
DA064222.1 | - | 2 | 328 |