Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001171092.1 | NP_001164563.1 | 2 | 2130 | |
NM_001171095.1 | NP_001164566.1 | 2 | 1899 | ||
NM_020384.3 | NP_065117.1 | 2 | 1965 | ||
GenBank mRNA | AK075371.1 | - | 2 | 1841 | |
AK075405.1 | - | 2 | 1937 |