Gene Transcripts

Gene Symbol
NR1I2
Entrez Gene ID
8856
Gene Name
nuclear receptor subfamily 1 group I member 2
Gene Aliases
BXR, ONR1, PAR, PAR1, PAR2, PARq, PRR, PXR, SAR, SXR
Chromosome Location
Chr. 3: 119780484 - 119818485 on Build GRCh38
UniGene ID
Hs.7303
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003889.3 NP_003880.3 9 3424
NM_022002.2 NP_071285.1 9 1750
NM_033013.2 NP_148934.1 9 3313
GenBank mRNA AF061056.1 - 9 1888
AF084644.1 - 9 1761
AF084645.1 - 9 1864
AJ009936.1 - 10 3425
AJ009937.1 - 10 3314
AK122990.1 - 6 1717
BC017304.2 - 9 1821 5087401

View Full Product Details