Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003827.3 | NP_003818.2 | 2 | 443 | |
NR_038456.1 | 2 | 443 | |||
XM_011527436.1 | XP_011525738.1 | 2 | 117 | ||
GenBank mRNA | AK098401.1 | - | 2 | 276 | |
AK292244.1 | - | 2 | 207 | ||
AK308633.1 | - | 3 | 333 | ||
AK310185.1 | - | 2 | 200 | ||
AY927609.1 | - | 2 | 225 | ||
BC001165.1 | - | 2 | 215 | 3533668 | |
BC007432.2 | - | 2 | 206 | 2962847 | |
BC028234.1 | - | 2 | 261 | 5245770 | |
BC091511.1 | - | 2 | 222 | 30512911 | |
BQ948081.1 | - | 2 | 425 | ||
BT019568.1 | - | 2 | 125 | ||
U39412.1 | - | 2 | 192 |