Gene Transcripts
- Gene Symbol
- MPDZ
- Entrez Gene ID
- 8777
- Gene Name
- multiple PDZ domain crumbs cell polarity complex component
- Gene Aliases
- HYC2, MUPP1
- Chromosome Location
- Chr. 9: 13105296 - 13279687 on Build GRCh38
- UniGene ID
- Hs.169378
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001261406.1 |
NP_001248335.1
|
4, 5 |
623 |
|
NM_001261407.1 |
NP_001248336.1
|
4, 5 |
623 |
|
NM_003829.4 |
NP_003820.2
|
4, 5 |
623 |
|
XM_005251622.4 |
XP_005251679.1
|
4, 5 |
572 |
|
XM_005251623.4 |
XP_005251680.1
|
4, 5 |
572 |
|
XM_006716885.3 |
XP_006716948.1
|
4, 5 |
621 |
|
XM_006716886.3 |
XP_006716949.1
|
5, 6 |
780 |
|
XM_006716887.3 |
XP_006716950.1
|
4, 5 |
724 |
|
XM_006716888.3 |
XP_006716951.1
|
4, 5 |
572 |
|
XM_006716889.3 |
XP_006716952.1
|
4, 5 |
575 |
|
XM_006716891.3 |
XP_006716954.1
|
4, 5 |
575 |
|
XM_017015252.1 |
XP_016870741.1
|
4, 5 |
572 |
|
XM_017015253.1 |
XP_016870742.1
|
4, 5 |
560 |
|
XM_017015254.1 |
XP_016870743.1
|
5, 6 |
783 |
|
XM_017015255.1 |
XP_016870744.1
|
4, 5 |
631 |
|
XM_017015256.1 |
XP_016870745.1
|
4, 5 |
572 |
|
XM_017015257.1 |
XP_016870746.1
|
4, 5 |
559 |
|
XM_017015258.1 |
XP_016870747.1
|
4, 5 |
572 |
|
XM_017015259.1 |
XP_016870748.1
|
4, 5 |
572 |
|
GenBank mRNA |
AB210041.1 |
- |
4, 5 |
624 |
|
AF093419.1 |
- |
3, 4 |
447 |
|
BC140793.1 |
- |
3, 4 |
447 |
9021661 |
BC144564.1 |
- |
3, 4 |
447 |
9053092 |
View Full Product Details