Gene Transcripts

Gene Symbol
THRSP
Entrez Gene ID
7069
Gene Name
thyroid hormone responsive
Gene Aliases
LPGP1, Lpgp, S14, SPOT14, THRP
Chromosome Location
Chr. 11: 78063861 - 78068357 on Build GRCh38
UniGene ID
Hs.591969
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003251.3 NP_003242.1 2 631
GenBank mRNA BC031989.1 - 2 614
BG674197.1 - 2 586

View Full Product Details