Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003253.2 | NP_003244.2 | 5 | 1243 | |
XM_005261037.1 | XP_005261094.1 | 4 | 1376 | ||
XM_005261038.2 | XP_005261095.1 | 3 | 1110 | ||
XM_005261040.2 | XP_005261097.1 | 4 | 1376 | ||
XM_011529711.1 | XP_011528013.1 | 5 | 1327 | ||
XM_011529712.1 | XP_011528014.1 | 5 | 1251 | ||
XM_011529713.1 | XP_011528015.1 | 5 | 1661 | ||
XM_017028447.1 | XP_016883936.1 | 5 | 1430 | ||
XM_017028448.1 | XP_016883937.1 | 5 | 1400 | ||
XM_017028449.1 | XP_016883938.1 | 4 | 1307 | ||
XM_017028450.1 | XP_016883939.1 | 4 | 1327 | ||
XM_017028451.1 | XP_016883940.1 | 4 | 1807 | ||
XM_017028452.1 | XP_016883941.1 | 6 | 1607 | ||
XM_017028453.1 | XP_016883942.1 | 4 | 1282 | ||
GenBank mRNA | AB209101.1 | - | 1 | 259 | |
BC117192.1 | - | 2 | 818 | 40125743 | |
BC117196.1 | - | 2 | 818 | 40125747 | |
BC143970.1 | - | 2 | 818 | ||
BC143980.1 | - | 2 | 818 | 9052498 | |
U16296.1 | - | 5 | 1244 |