Gene Transcripts

Gene Symbol
ZNF282
Entrez Gene ID
8427
Gene Name
zinc finger protein 282
Gene Aliases
HUB1
Chromosome Location
Chr. 7: 149195462 - 149226248 on Build GRCh38
UniGene ID
Hs.729056
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001303481.1 NP_001290410.1 9 2294
NM_003575.3 NP_003566.1 8 2551
XM_006716151.3 XP_006716214.1 8 2548
GenBank mRNA BC031686.1 - 4 1306
BC041704.1 - 8 2278
BC055287.1 - 8 2343
BC073805.1 - 8 2444 6645861
D30612.1 - 8 2564

View Full Product Details