Gene Transcripts

Gene Symbol
CDK13
Entrez Gene ID
8621
Gene Name
cyclin dependent kinase 13
Gene Aliases
CDC2L, CDC2L5, CHED, hCDK13
Chromosome Location
Chr. 7: 39949501 - 40097134 on Build GRCh38
UniGene ID
Hs.233552
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003718.4 NP_003709.3 2 1528
NM_031267.3 NP_112557.2 2 1528
XM_011515597.2 XP_011513899.1 2 2387
XM_017012750.1 XP_016868239.1 2 2387
XM_017012751.1 XP_016868240.1 2 2387
GenBank mRNA AJ297709.1 - 2 1378
AJ297710.1 - 2 1378
AK092534.1 - 2 1528
BC080601.1 - 2 1851
M80629.1 - 2 175

View Full Product Details