Gene Transcripts

Gene Symbol
CDK13
Entrez Gene ID
8621
Gene Name
cyclin dependent kinase 13
Gene Aliases
CDC2L, CDC2L5, CHED, hCDK13
Chromosome Location
Chr. 7: 39949501 - 40097134 on Build GRCh38
UniGene ID
Hs.233552
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003718.4 NP_003709.3 2 2015
NM_031267.3 NP_112557.2 2 2015
XM_011515597.2 XP_011513899.1 2 2874
XM_017012750.1 XP_016868239.1 2 2874
XM_017012751.1 XP_016868240.1 2 2874
GenBank mRNA AJ297709.1 - 2 1865
AJ297710.1 - 2 1865
AK092534.1 - 2 2015
BC001274.1 - 1 285
BC080601.1 - 2 2338
M80629.1 - 2 662

View Full Product Details