Gene Transcripts
- Gene Symbol
- ZBTB16
- Entrez Gene ID
- 7704
- Gene Name
- zinc finger and BTB domain containing 16
- Gene Aliases
- PLZF, ZNF145
- Chromosome Location
- Chr. 11: 114059576 - 114256765 on Build GRCh38
- UniGene ID
- Hs.591945
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001018011.1 |
NP_001018011.1
|
4, 5 |
1562 |
|
NM_006006.4 |
NP_005997.2
|
4, 5 |
1717 |
|
XM_017018257.1 |
XP_016873746.1
|
4, 5 |
1650 |
|
XM_017018258.1 |
XP_016873747.1
|
4, 5 |
1805 |
|
XM_017018259.1 |
XP_016873748.1
|
4, 5 |
2157 |
|
GenBank mRNA |
BC026902.1 |
- |
4, 5 |
1621 |
|
BC029812.1 |
- |
4, 5 |
1699 |
5175961 |
BX648973.1 |
- |
4, 5 |
1834 |
|
KU178400.1 |
- |
3, 4 |
1453 |
|
Z19002.1 |
- |
3, 4 |
1528 |
|
View Full Product Details